missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VCAM-1 (CD106) Monoclonal Antibody (6C7.1)
Rat Monoclonal Antibody
Brand: Invitrogen™ MA549138
This item is not returnable.
View return policy
Description
VCAM-1 (CD106) Monoclonal Antibody for ICC/IF, IHC (F), Flow, IP, Functional Assay
VCAM-1 is a cell surface sialoglycoprotein expressed by cytokine activated endothelium. The protein has a number of functions including the regulation of leukocyte migration, leukocyte-endothelial cell adhesion and signal transduction and may play a role in a number of inflammatory diseases.
Specifications
| VCAM-1 (CD106) | |
| Monoclonal | |
| 100 μg/mL | |
| PBS with 0.1% BSA and no preservative | |
| P29533 | |
| VCAM1 | |
| Mouse endothelial cells. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Functional Assay, Immunohistochemistry (Frozen), Immunoprecipitation, Immunocytochemistry | |
| 6C7.1 | |
| Unconjugated | |
| VCAM1 | |
| adhesion molecule VCAM-1; CD106; CD106 antigen; DKFZp779G2333; INCAM100; INCAM-100; MGC99561; sCD106; soluble CD106; soluble VCAM 1; sVCAM 1; vascular cell adhesion molecule; vascular cell adhesion molecule 1; vascular cell adhesion molecule-1; vascular cell adhesion protein 1; V-CAM 1; Vcam1; Vcam-1; VCAM1B | |
| Rat | |
| Protein G | |
| RUO | |
| 22329 | |
| 4°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction