Learn More
Invitrogen™ VE-cadherin Recombinant Rabbit Monoclonal Antibody (9W5E3)

Rabbit Recombinant Monoclonal Antibody
176.00€ - 436.00€
Specifications
| Antigen | VE-cadherin |
|---|---|
| Clone | 9W5E3 |
| Concentration | 0.13 mg/mL |
| Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30283575
|
Invitrogen™
MA557859 |
20 μL |
176.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30283805
![]() |
Invitrogen™
MA557860 |
100 μL |
436.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Immunogen sequence: DWIWNQMHID EEKNTSLPHH VGKIKSSVSR KNAKYLLKGE YVGKVFRVDA ETGDVFAIER LDRENISEYH LTAVIVDKDT GENLETPSSF TIKVHDVNDN WPVFTHRLFN ASVPESSAVG TSVISVTAVD ADDPTVGDHA SVMYQILKGK EYFAIDNSGR IITITKSLDR EKQARYEIVV EARDAQGLRG DSGTATVLVT LQDINDNFPF FTQTKYTFVV PEDTRVGTSV GSLFVEDPDE PQNRMTKYSI LRGDYQDAFT IETNPAHNEG IIKPMKPLDY EYIQQYSFIV EATDPTIDLR YMSPPAGNRA QVIINITDVD EPPIFQQPFY HFQLKENQKK PLIGTVLAMD PDAARHSIGY SIRRTSDKGQ FFRVTKKGDI YNEKELDREV YPWYNLTVEA KELDSTGTPT GKESIVQVHI EVLDENDNAP EFAKPYQPKV CENAVHGQLV LQISAIDKDI TPRNVKFKFI LNTENNFTLT DNHDNTANIT VKYGQFDREH TKVHFLPVVI SDNGMPSRTG TSTLTVAVCK CNEQGEFTFC EDMAAQ.
VE-cadherin is a member of the cadherin superfamily that is located in a six-cadherin cluster in a region on the long arm of chromosome 16 and is involved in loss of heterozygosity events in breast and prostate cancer. VE-cadherin protein is a calcium-dependent cell-cell adhesion glycoprotein comprised of five extracellular cadherin repeats, a transmembrane region and a highly conserved cytoplasmic tail. Functioning as a classic cadherin by imparting to cells the ability to adhere in a homophilic manner, VE-cadherin may play an important role in endothelial cell biology through control of the cohesion and organization of the intercellular junctions. An alternative splice variant has been described but the full length sequence of VE-cadherin has not been determined.Specifications
| VE-cadherin | |
| 0.13 mg/mL | |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| P33151 | |
| 1003 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 48-593 of human VE Cadherin (NP_001786.2). | |
| Antibody |
| 9W5E3 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human | |
| CDH5 | |
| 7B4; 7B4 antigen; 7B4/cadherin-5; AA408225; cadherin 5; cadherin 5, type 2 (vascular endothelium); cadherin 5, type 2, VE-cadherin (vascular endothelium); cadherin 5, type 2, VE-cadherin (vascular epithelium); cadherin-5; cadherin-5; cd144 antigen; VE cadherin; Cd144; cd144 antigen; CDH5; cell-cell adhesion protein; endothelial-specific cadherin; vascular endothelial cadherin; vascular endothelial cadherin precursor; Vec; VEcad; VE-Cad; VE-cadherin; VECD | |
| CDH5 | |
| Primary | |
| Protein A |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.