Learn More
Asecos™ Ventilation Unit Box
Product Code. 12831183 Shop All asecos Products
Change view
Click to view available options
Dimensions (L x W x H):
410 x 430 x 300 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W x H) Unit Size
12831183 410 x 430 x 300 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12831183 Supplier Asecos™ Supplier No. EP.L.53049
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
Ventilation Unit Box for Corrosive Products. Soundproof ventilation box with integrated filtration.
Specifications
For Use With (Application) Corrosive Products
Product Type Ventilation Unit Box
Dimensions (L x W x H) 410 x 430 x 300 mm
Material Steel construction, epoxy coating