Learn More
Elga™ Pre-Treatment Cartridge
Product Code. 11994857
Change view
Click to view available options
For Use With (Equipment):
Purelab™ Option-S Series Water Purification Systems
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
11994857 Purelab™ Option-S Series Water Purification Systems Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11994857 Supplier Elga™ Supplier No. LC140
  • ONLINE EXCLUSIVE PRICE
    570.00€
    330.67€ / Pack of 1
    Save 239.33€
    41% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
Protects RO membranes by removing particulates and chlorine.
  • Pretreatment cartridge
  • Helps to produce general laboratory grade water at better than single distilled quality
  • Applications include glassware washing/rinsing, feed to ultra pure water systems, reagent make-up/dilution, buffer and media preparation, general chemistry, qualitative analyses and feed to environmental cabinets
Specifications
For Use With (Application) Glassware washing/rinsing, feed to ultra pure water systems, reagent make-up/dilution, buffer and media preparation, general chemistry, qualitative analyses and feed to environmental cabinets
For Use With (Equipment) Purelab™ Option-S Series Water Purification Systems