Learn More
Veolia Water Systems™ UV Lamp
Product Code. 11954897
Change view
Click to view available options
For Use With (Equipment):
Bactericide and to break down and photo-oxidize organic contaminants to polar or ionized species
PURELAB Option-R
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
11954897 PURELAB Option-R Each
12013283 Bactericide and to break down and photo-oxidize organic contaminants to polar or ionized species Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11954897 Supplier Veolia Water Systems™ Supplier No. LC105
  • ONLINE EXCLUSIVE PRICE
    493.50€
    405.44€ / Pack of 1
    Save 88.06€
    17% Off
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
Rich, robust UV lamp that features UV light acting as a bactericide to break down and photo-oxidize organic contaminants to polar or ionized species.
  • UV light is used as a bactericide and to break down and photo-oxidise organic contaminants to polar or ionized species
  • The UV bactericidal action damages DNA and RNA polymerase at low doses thereby preventing replication
Specifications
For Use With (Application) Break down and photo-oxidize organic contaminants to polar or ionized species
For Use With (Equipment) PURELAB Option-R