Learn More
Veolia Water Systems Veolia Water Systems™ Water Storage System
Product Code. 14865211
Change view
Click to view available options
Capacity (Metric):
30 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
14865211 30 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 14865211 Supplier Veolia Water Systems Supplier No. LA758
  • ONLINE EXCLUSIVE PRICE
    2667.00€
    2115.34€ / Pack of 1
    Save 551.66€
    20% Off
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Robust, reliable water storage system with 30L capacity.
  • 30 liter storage resevoir
  • Storage Water System
Specifications
Capacity (Metric) 30 L