Learn More
Emma Safety Footwear Vera Safety Shoes, Red
Product Code. 15968511
Change view
Click to view available options
Size:
37
38
39
40
41
42
43
44
45
46
47
48
49
Unit Size:
1 pair
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15968511 37 1 pair
15978511 38 1 pair
15988511 39 1 pair
15998511 40 1 pair
15908521 41 1 pair
15918521 42 1 pair
15928521 43 1 pair
15938521 44 1 pair
15948521 45 1 pair
15958521 46 1 pair
15968521 47 1 pair
15978521 48 1 pair
15988521 49 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Product Code. 15968511 Supplier Emma Safety Footwear Supplier No. MM50652437
  • ONLINE EXCLUSIVE PRICE
    178.92€
    87.99€ / 1 pairs
    Save 90.93€
    50% Off
Estimated Shipment: 28-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
An easy shoe for wipe clean hygiene. The upper is fully washable unique microfiber and features Hydro-Tec™ Sanitized Silver for dry, fresh feet. The protective toe cap prevents staining and is lubricant and fat resistant. Developed for use on hard tiled floors and other easy-to-clean floors.
  • S2-Safety shoe for wet working conditions
  • Microfiber construction
  • GripForce™ Duo Heat Resistant Outsole
  • Chrome-free lining
  • Sanitized
Specifications
Certifications/Compliance EN ISO 20345:2011
Closure Type Elastic
Color White
Design Vera, Red
Liner Material Hydro-Tec™ Sanitized Silver
Material Microfiber
Size 37
Toe Material Steel