Learn More
Unity Lab Services™ Verification Report Autoclave
Product Code. 12568526
Change view
Click to view available options
For Use With (Equipment):
Autoclaves
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
12568526 Autoclaves Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12568526 Supplier Unity Lab Services™ Supplier No. UMVP9323
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
ULS offers a full range of support services and programs for all instruments and supplies, regardless of manufacturer. Maximize your technology investments by anticipating and avoiding problems before they occur.
  • Pressure and temperature control at 1.05 bar (121°C) and 2.04 bar (134°C)
  • Service performed with a COFRAC pressure gauge
  • Provision of a verification report
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Autoclaves
Duration 1 Day
Product Type Verification Report