missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse B7-2/CD86 (NP_062261) VersaClone cDNA

Product Code. 18783379
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18783379 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18783379 Supplier R&D Systems™ Supplier No. RDC1181
  • ONLINE EXCLUSIVE PRICE
    341.00€
    322.35€ / 10µg
    Save 18.65€
    5% Off
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

B7-1 and B7-2, together with their receptors CD28 and CTLA4, constitute one of the dominant costimulatory pathways that regulate T and B cell responses.

The Mouse B7-2/CD86 (NP_062261) VersaClone cDNA clone contains the complete ORF for B7-2/CD86, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The B7-2/CD86 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.

Order Info

Ships at ambient temperature

TRUSTED_SUSTAINABILITY

Specifications

Gene B7-2/CD86
Accession Number NP_062261
Concentration 10 μg at 0.2 μg/μL
Formulation cDNA is provided in 10mM Tris-Cl, pH 8.5
Insert Type 943bp
Gene ID (Entrez) 942
Gene Alias Activation B7-2 antigen, B70, B72, B7-2, B7-2 antigen, B-lymphocyte activation antigen B7-2, BU63, CD28 antigen ligand 2, CD28LG2B7-2 antigen), CD86, CD86 antigen, CD86 molecule, CTLA-4 counter-receptor B7.2, FUN-1, LAB72, MGC34413, T-lymphocyte activation antigen CD86
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Mouse
Quantity 10 μg

For Reasearch Use Only; Not for Use in Humans

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.