Learn More
3M™ Versaflo™ Intrinsically Safe Powered Air Turbo Starter Kit, TR-819UK
Description
- Convenient ready-to-use kit
- Approved for use with a wide range of Versaflo™ Soft Headtops S Series and Rigid Headtops M Series
- Includes a rechargeable standard capacity lithium Ion battery
- Headtop not included
- CE approved
Specifications
Specifications
| Battery Type | Lithium-ion |
| Certifications/Compliance | CE Approved |
| Product Type | PAPR Systems |
| Quantity | 1/Cs. |