Learn More
3M™ Versaflo™ Premium S-Series Hood and Hard Hat Adapter Assembly, S-960
Product Code. 31297041 Shop All 3M Products
Change view
Click to view available options
Product Type:
Hood and Hard Hat Adapter Assembly
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type
31297041 Hood and Hard Hat Adapter Assembly
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 31297041 Supplier 3M™ Supplier No. 7100308819
  • ONLINE EXCLUSIVE PRICE
    129.00€
    83.89€ / Pack of 1
    Save 45.11€
    34% Off
Estimated Shipment: 09-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Designed to fit the 3M™ SecureFit™ Hard Hat H-700 Series and the 3M™ Hard Hat G3000 Series.
  • Compatible with 3M™ Versaflo™ S-Series Premium Hoods and 3M™ Versaflo™ Powered and Supplied Air Respirator Systems.
  • Provides integrated head, face, shoulder, and respiratory protection from dust and spray.
  • Helps direct airflow to the front of the hood for improved comfort and visibility.
  • Enables easy integration of head and respiratory protection for enhanced worker safety.
  • Lightweight and durable design for comfortable, long-term use in industrial environments.
TRUSTED_SUSTAINABILITY
Specifications
Color Black
Disposable Single-use
Material Plastic
Product Type Hood and Hard Hat Adapter Assembly
Size One Size