Learn More
3M™ Versaflo™ S-Series Hood Assembly with Sealed Seams Inner Collar and Helmet, S-865E
Product Code. 31297045 Shop All 3M Products
Change view
Click to view available options
Product Type:
PAPR Assembly with Hood
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type
31297045 PAPR Assembly with Hood
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 31297045 Supplier 3M™ Supplier No. 7100319508
  • ONLINE EXCLUSIVE PRICE
    398.00€
    260.43€ / Pack of 1
    Save 137.57€
    34% Off
Estimated Shipment: 16-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Offer a variety of options for faceseals, collars, shrouds, and suspensions, allowing you to choose the respiratory protection that provides the most value and comfort for your team.
  • Lightweight, versatile and easy to maintain
  • Help protect your workers from a variety of contaminants while maintaining comfort
  • Available in fully assembled integrated hoods for convenience
  • Reusable suspension options offer cost savings
  • Designed to work for your needs
TRUSTED_SUSTAINABILITY
Specifications
Color Gray
Disposable Single-use
Filter Type Combination
Material Plastic
Product Type PAPR Assembly with Hood
Size One Size
With Exhalation Valve No