Learn More
Thermo Scientific™ Versette™ Accessories
Product Code. 11664922 Shop All Thermo Scientific Products
Change view
Click to view available options
Description:
384-Channel Tip Wash Station, Tall Height
96-Channel Tip Wash Station, Tall Height
Pump module; Optional accessory, used for tip washing/reagent replenishing; 12.7 kg (28 lbs.)
Serial dilute magazine 384/30&100μL (16/24)
Serial dilute magazine 96/30μL (8/12) for use with short 30μL tips
Serial dilute magazine 96/300Glc muL (8/12)
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Unit Size
11664922 96-Channel Tip Wash Station, Tall Height Each
15493770 Pump module; Optional accessory, used for tip washing/reagent replenishing; 12.7 kg (28 lbs.) Each
15423780 Serial dilute magazine 384/30&100μL (16/24) Each
13555780 Serial dilute magazine 96/300Glc muL (8/12) Each
15403780 Serial dilute magazine 96/30μL (8/12) for use with short 30μL tips Each
11654922 384-Channel Tip Wash Station, Tall Height Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 11664922 Supplier Thermo Scientific™ Supplier No. 6500596TTW
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Use Thermo Scientific™ Versette™ Accessories with the Versette™ Automated Liquid Handling System.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Tip Wash Station
Description 96-Channel Tip Wash Station, Tall Height
Channels 96
What accessories are available for the Versette Automated Liquid Handler?
Here are the accessories available for the Versette Automated Liquid Handler:
- Three pipetting heads (volume range 0.5 µL - 300 µL)
- Peristaltic pump including non-contact reservoir fill
- Tip wash stations for 96/384-well formats
- Four serial dilution magazines for 96/384-well applications
What is the warranty on the Versette Automated Liquid Handler, including components, e.g., pipetting heads, pump, and tip wash station?
The warranty is 12 months from the date of shipment.
Can reservoirs be automatically filled with the Versette Automated Liquid Handler?
Yes, a non-contact (N/C) reservoir fill module is included as an accessory with the purchase of a peristaltic pump (Cat. No. 650-04-PUMP).