missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vesicular Stomatitis Indiana Virus Glycoprotein Antibody (110) - (San Juan), Novus Biologicals™
Rabbit Monoclonal Antibody
214.00€ - 410.00€
Specifications
| Antigen | Vesicular Stomatitis Indiana Virus Glycoprotein |
|---|---|
| Clone | 110 |
| Dilution | ELISA 1:5000-1:10000 |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30239500
|
Novus Biologicals
NBP3-40579-100ul |
100 μL |
410.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30238495
|
Novus Biologicals
NBP3-40579-20ul |
20 μL |
214.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Vesicular Stomatitis Indiana Virus Glycoprotein Monoclonal antibody specifically detects Vesicular Stomatitis Indiana Virus Glycoprotein in Virus samples. It is validated for ELISASpecifications
| Vesicular Stomatitis Indiana Virus Glycoprotein | |
| ELISA 1:5000-1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| 0.2 μm filtered solution in PBS | |
| IgG | |
| Protein A purified |
| 110 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Virus | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Vesicular Stomatitis Indiana Virus Glycoprotein - (San Juan) (RefSeq#: NP_041715.1; Met1-Ser467). HEK293 used as expression system. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title