missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VGLUT1 Antibody (CL2754) [Alexa Fluor™ 350], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-46627AF350
This item is not returnable.
View return policy
Description
VGLUT1 Monoclonal antibody specifically detects VGLUT1 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| VGLUT1 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| BNPIbrain-specific Na-dependent inorganic phosphate cotransporter, Brain-specific Na(+)-dependent inorganic phosphate cotransporter, member 7, solute carrier family 17 (sodium-dependent inorganic phosphate cotransporter), Solute carrier family 17 member 7, VGluT1, VGLUT1vesicular glutamate transporter 1 | |
| This antibody was developed using a recombinant protein derived from Q9P2U7, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| ABC Transporters, Cellular Signaling, Neuronal Cell Markers, Neuroscience | |
| 57030 | |
| Store at 4⪚C in the dark. | |
| IgG2b |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL2754 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction