Learn More
beta Tubulin 1,2 Monoclonal Antibody (JDR.3B8), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA125057
This item is not returnable.
View return policy
Description
Recommended positive controls: chicken fibroblasts, chicken brain. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
beta Tubulin 1,2 Monoclonal antibody specifically detects beta Tubulin 1,2 in Bovine, Chicken, Human, Mouse, Porcine, Rat samples. It is validated for Immunocytochemistry, Western Blot.
Specifications
| beta Tubulin 1,2 | |
| Monoclonal | |
| Conc. Not Determined | |
| Ascites with 15mM sodium azide | |
| A2AQ07, P07437, P09207, P09244, P85108, Q13885, Q2KJD0, Q767L7, Q7TMM9, Q9H4B7 | |
| TUBB, TUBB1, TUBB2A | |
| A chemically synthesized peptide corresponding to the carboxyl-terminal sequence of human beta-tubulin isotype II coupled to BSA | |
| RUO | |
| 100134983, 100295973, 203068, 22151, 396254, 396427, 498736, 541271, 545486, 615087, 679312, 7280, 733686, 768337, 81027 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Western Blot | |
| JDR.3B8 | |
| Unconjugated | |
| TUBB | |
| 2810484G07Rik; beta 5-tubulin; beta Ib tubulin; beta-tubulin; beta-Tubulin 1; beta-Tubulin 2; beta-tubulin class-VI; c(beta)7 tubulin; CDCBM5; CDCBM6; class IIa beta-tubulin; CSCSC1; M(beta)1; M(beta)2; M40; MGC128415 protein; OK/SW-cl.56; tbb1; TUBB; TUBB1; Tubb2; Tubb2a; tubb2a {ECO:0000250; TUBB5; tubulin beta; tubulin beta 1 class VI; tubulin beta 2A class IIa; tubulin beta 4'; Tubulin beta chain; tubulin beta class I; Tubulin beta class IIa; Tubulin beta-1 chain; tubulin beta-1 chain-like; tubulin beta-2A chain; tubulin beta-2B chain; tubulin beta-5 chain; Tubulin beta-6 chain; tubulin beta-7 chain; tubulin, beta 1 class VI; tubulin, beta 2A; tubulin, beta 2A class IIa; tubulin, beta 3; tubulin, beta class I; tubulin, beta family 1; tubulin, beta polypeptide; tubulin, beta polypeptide 2; UniProtKB:Q7TMM9}; unnamed protein product | |
| Mouse | |
| 100 μL | |
| Primary | |
| Bovine, Chicken, Human, Mouse, Pig, Rat | |
| Antibody | |
| IgG2b |