Learn More
RETSCH CryoMill
Product Code. 13454886 Shop All Retsch Products
Change view
Click to view available options
Voltage:
100/240 V
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Voltage unitSize
13454886 100/240 V Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13454886 Supplier RETSCH Supplier No. 20.749.0001
Add to basket
Add to basket
Designed for cryogenic grinding. Retsch™ CryoMills provide powerful impact ball milling which results in a perfect grinding efficiency.
  • Powerful cryogenic grinding by impact and friction, up to 30 Hz
  • 3 different grinding modes (cryogenic, dry or wet at ambient temperature)
  • Closed LN2-system (autofill) for enhanced safety, avoids any contact of the user with LN2
  • Screw-top grinding jars for convenient, leak-proof operation
  • Wide range of accessories including various LN2 feeding systems, jar and ball sizes, adapter racks, materials
  • Low LN2-consumption
  • Clearly structured user interface, memory for 9 SOPs
  • Programmable cooling and grinding cycles (10 sec. to 99 min.)
  • Ceramic jar available
Specifications
Product Type CryoMill
Certifications/Compliance CE Certified
Dimensions (L x W x H) 675 x 385 x 370 mm
Electrical Requirements 100/240 V, 50/60 Hz
For Use With (Application) Agriculture, biology, chemistry/Plastics, construction materials, engineering/electronics, environment/recycling, food, geology/metallurgy, glass/ceramics, medicine/pharmaceuticals
Height (Metric) 370 mm
Hertz 50/60 Hz
IP Rating IP30
Length (Metric) 675 mm
Phase 1-phase
Voltage 100/240 V
Width (Metric) 385 mm