Learn More
ETNK2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538807
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a member of choline/ethanolamine kinase family which catalyzes the first step of phosphatidylethanolamine (PtdEtn) biosynthesis via the cytidine diphosphate (CDP) ethanolamine pathway. Alternative splicing results in multiple transcript variants. [provided by RefSeq, Jul 2014].
Specifications
| ETNK2 | |
| Polyclonal | |
| Unconjugated | |
| ETNK2 | |
| EKI2, HMFT1716, EKI 2, ethanolamine kinase-like protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 214253, 55224 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9NVF9, A7MCT6 | |
| ETNK2 | |
| A synthetic peptide derived from the internal region of human EKI2 | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |