Learn More
Invitrogen™ SRSF7 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560537
This item is not returnable.
View return policy
Description
Immunogen sequence: SRVRVELSTG MPRRSRFDRP PARRPFDPND RCYECGE Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
PMVK is a peroxisomal enzyme that catalyzes the conversion of mevalonate 5-phosphate into mevalonate 5-diphosphate as the fifth reaction of the cholesterol biosynthetic pathway.
Specifications
| SRSF7 | |
| Polyclonal | |
| Unconjugated | |
| Srsf7 | |
| 35kDa; 9430065L19Rik; 9G8; AAG3; aging-associated protein 3; hypothetical protein LOC507066; NX-96; serine and arginine rich splicing factor 7; serine/arginine-rich splicing factor 7; serine/arginine-rich splicing factor 7-like protein; SFRS7; Splicing factor 9G8; splicing factor, arginine/serine-rich 7; splicing factor, arginine/serine-rich 7, 35kDa; SR splicing factor 7; SRSF7 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6432 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q16629 | |
| Srsf7 | |
| Recombinant protein corresponding to Human SRSF7. Recombinant protein control fragment (Product #RP-99638). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |