Learn More
Invitrogen™ KCC1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA559444
This item is not returnable.
View return policy
Description
Immunogen sequence: MPHFTVVPVD GPRRGDYDNL EGLSWVDYGE RAELDDSDGH GNHRESSPFL SPLEASRGID YYDRNL Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 91%.
SLC12A4 mediates electroneutral potassium-chloride cotransport when activated by cell swelling. SLC12A4 may contribute to cell volume homeostasis in single cells.
Specifications
| KCC1 | |
| Polyclonal | |
| Unconjugated | |
| SLC12A4 | |
| AW546649; CTC-479C5.17; Electroneutral potassium-chloride cotransporter 1; erythroid K-Cl cotransporter 1; FLJ40489; Furosemide-sensitive K-Cl cotransporter; hKCC1; Kcc1; K-Cl cotransporter; K-Cl cotransporter 1; K-Cl cotransporter 4; K-Cl cotransporter KCC1; K-Cl Co-transporter-1; mKCC1; potassium chloride cotransporter SLC12A4; potassium/chloride cotransporter 1; potassium-chloride cotransporter-1; RBCKCC1; RBKCC1; rKCC1; Slc12a4; solute carrier family 12 (potassium/chloride transporter), member 4; solute carrier family 12 (potassium/chloride transporters), member 4; solute carrier family 12 member 4; solute carrier family 12, member 4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6560 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UP95 | |
| SLC12A4 | |
| Recombinant protein corresponding to Human KCC1. Recombinant protein control fragment (Product #RP-98562). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |