Learn More
CD4 Mouse anti-Baboon,Cynomolgus,Rhesus Ma,Human, BB700, Clone: L200, BD Horizon™
Mouse Monoclonal Antibody
Brand: BD Biosciences 566479
This item is not returnable.
View return policy
Description
CD4 Monoclonal antibody specifically detects CD4 in Baboon,Cynomolgus,Rhesus Ma,Human samples. It is validated for Flow Cytometry.
Specifications
| CD4 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Mouse | |
| RUO | |
| Baboon, Cynomolgus, Rhesus Macaque, Human | |
| Purified |
| Flow Cytometry | |
| L200 | |
| Brilliant Blue 700 (BB700) | |
| 0.1 mg | |
| Primary | |
| Refrigerate (2 to 8°C) | |
| IgG1 κ |