Learn More
VSX2 Sheep anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Sheep polyclonal antibody raised against partial recombinant VSX2.
Brand: Abnova PAB9842.250ug
This item is not returnable.
View return policy
Specifications
| VSX2 | |
| Polyclonal | |
| Sheep polyclonal antibody raised against partial recombinant VSX2. | |
| In PBS (0.08% sodium azide) | |
| CHX10/HOX10/MCOP2/MCOPCB3/RET1 | |
| Sheep | |
| 250 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
| Immunohistochemistry (Frozen), Western Blot | |
| Unconjugated | |
| Western Blot (0.5-1.0 ug/mL) The optimal working dilution should be determined by the end user. | |
| VSX2 | |
| VSX2 | |
| Recombinant GST fusion protein corresponding to amino acids 264-361 of human VSX2. | |
| RUO | |
| 338917 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |