Learn More
Invitrogen™ FBF1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5117947
This item is not returnable.
View return policy
Description
FBF1 Polyclonal Antibody for Western Blot, IP
Keratin-binding protein required for epithelial cell polarization. Involved in apical junction complex (AJC) assembly via its interaction with PARD3. Required for ciliogenesis.
Specifications
| FBF1 | |
| Polyclonal | |
| PBS with 0.03% ProClin 300; pH 7 | |
| Q8TES7 | |
| FBF1 | |
| E. coli-derived Human FBF1 fragment. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| FBF1 | |
| 1110033G01Rik; 5930413D20; Alb; albatross; Fas (TNFRSF6) binding factor 1; Fas binding factor 1; Fas binding protein 1; fas-binding factor 1; Fbf1; FBF-1; KIAA1863; likely ortholog of H. sapiens Fas (TNFRSF6) binding factor 1 (FBF1); mKIAA1863; Protein albatross | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 85302 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |