Learn More
SARS-CoV-2 Spike Antibody (CR3022), DyLight 405 - Chimeric, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP2-90979V
Product Code. 18080197
  • 604.00€ / 0.10mL
Description
SARS-CoV-2 Spike Monoclonal antibody specifically detects SARS-CoV-2 Spike in SARS-CoV-2, SARS-CoV samples. It is validated for ELISA, Immunocytochemistry, Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SARS-CoV-2 Spike
Monoclonal
DyLight 405
2019-nCoV S Protein, 2019-nCoV Spike, COVID-19 Spike, Human coronavirus spike glycoprotein, SARS-CoV-2, SARS-COV-2 S protein, SARS-COV-2 Spike glycoprotein, SARSCOV2 Spike protein, Severe Acute Respiratory Syndrome Coronavirus 2 Spike Protein, Spike glycoprotein
Rabbit
Protein A purified
RUO
This antibody binds to both SARS-CoV and SARS-CoV-2 with high affinity (PMID: 16796401 & 32065055). It binds the amino acids 318-510 in the S1 domain of the SARS-CoV Spike protein as well as SARS-CoV-2 (COVID-19) Spike protein. The antibody also binds to P462L-substituted S318-510 fragments of the SARS spike protein. The binding epitope is only accessible in the "open" confromation of the spike protein (Joyce et al. 2020). NBP2-90979 cross-reacts with spike protein of Omicron and Delta variants.
Store at 4°C in the dark.
IgG κ
ELISA
CR3022
50mM Sodium Borate
S
The original monoclonal antibody was generated by sequencing peripheral blood lymphocytes of a patient exposed to the SARS-CoV.
0.1 mL
Primary
SARS-CoV