Learn More
GAGE1 Antibody (4G6), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00002543-M04
This item is not returnable.
View return policy
Description
GAGE1 Monoclonal antibody specifically detects GAGE1 in Human samples. It is validated for Western Blot, ELISA, ELISA
Specifications
| GAGE1 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| Antigen MZ2-F, Cancer/testis antigen 4.1, G antigen 1, GAGE-1, member 1, MGC33825, MZ2-F antigen | |
| GAGE1 (NP_001459.2, 1 a.a. ~ 139 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MSWRGRSTYYWPRPRRYVQPPEMIGPMRPEQFSDEVEPATPEEGEPATQRQDPAAAQEGEDEGASAGQGPKPEADSQEQGHPQTGCECEDGPDGQEMDPPNPEEVKTPEEEMRSHYVAQTGILWLLMNNCFLNLSPRKP | |
| 0.1 mg | |
| Cancer | |
| 2543 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Western Blot, ELISA, Sandwich ELISA | |
| 4G6 | |
| Western Blot 1:100-1:2000, ELISA, Sandwich ELISA 1:100-1:2000 | |
| NP_001459.2 | |
| Mouse | |
| IgG purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |