Learn More
Cathepsin Z Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06499IR
This item is not returnable.
View return policy
Description
Cathepsin Z Polyclonal antibody specifically detects Cathepsin Z in Human samples. It is validated for Immunofluorescence
Specifications
| Cathepsin Z | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunofluorescence | |
| DyLight 755 | |
| Cathepsin P, Cathepsin X, cathepsin Z, CTSX, EC 3.4.18.1, FLJ17088, preprocathepsin P | |
| Produced in rabbits immunized with E. coli-derived Human Cathepsin Z fragment. | |
| 0.1 mL | |
| Cancer | |
| 1522 | |
| Store at 4°C in the dark. | |
| IgG |