Learn More
CA19-9/Sialyl Lewis A Antibody (CA19.9/1390R), Janelia Fluor™ 646, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-54585JF646
This item is not returnable.
View return policy
Description
CA19-9/Sialyl Lewis A Monoclonal antibody specifically detects CA19-9/Sialyl Lewis A in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| CA19-9/Sialyl Lewis A | |
| Monoclonal | |
| Janelia Fluor 646 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CA19.9/1390R | |
| 50 mM Sodium Borate | |
| Purified human CA19-9/Sialyl Lewis A protein | |
| 0.1 mL | |
| Tumor Biomarkers | |
| Human | |
| Purified |