Learn More
Pancreatic trypsin inhibitor Antibody (18), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-23588MFV610
Product Code. 18474416
  • 610.00€ / 0.10mL
Description
Pancreatic trypsin inhibitor Monoclonal antibody specifically detects Pancreatic trypsin inhibitor in Bovine samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Pancreatic trypsin inhibitor
Monoclonal
mFluor Violet 610 SE
50mM Sodium Borate
Mouse
Protein A or G purified
RUO
404172
Store at 4C in the dark.
IgG1 κ
Western Blot, ELISA
18
Western Blot, ELISA
Aprotinin, Basic protease inhibitor, BPI, BPTI, PTI, PTI pancreatic trypsin inhibitor
Aprotinin from bovine lung.
0.1 mL
Primary
Bovine