Learn More
Coagulation Factor X Antibody (MM0192-10A10), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12198PE
This item is not returnable.
View return policy
Description
Coagulation Factor X Monoclonal antibody specifically detects Coagulation Factor X in Human samples. It is validated for Western Blot
Specifications
| Coagulation Factor X | |
| Monoclonal | |
| PE | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 2159 | |
| Store at 4C in the dark. | |
| IgG2 |
| Western Blot | |
| MM0192-10A10 | |
| Western Blot | |
| coagulation factor X, EC 3.4.21, EC 3.4.21.6, factor Xa, FX, FXA, prothrombinase, Stuart factor, Stuart-Prower factor | |
| Human recombinant Coagulation Factor X | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |