Learn More
FGF-21 Antibody (461804), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB25372MFV500
Product Code. 18546687
  • 652.00€ / 0.10mL
Description
FGF-21 Monoclonal antibody specifically detects FGF-21 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
FGF-21
Monoclonal
mFluor Violet 500 SE
50mM Sodium Borate
Mouse
0.1 mL
Angiogenesis, Biologically Active Proteins, Cancer
26291
Store at 4C in the dark.
Flow Cytometry, Immunocytochemistry, CyTOF
461804
Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready
FGF-21, fibroblast growth factor 21
E. coli-derived recombinant human FGF-21, His29-Ser209, Accession # Q9NSA1
RUO
Primary
Human
IgG2b