Learn More
HIBCH Antibody (OTI3H5), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71796CL1
This item is not returnable.
View return policy
Description
HIBCH Monoclonal antibody specifically detects HIBCH in Canine,Human samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,CyTOF
Specifications
| HIBCH | |
| Monoclonal | |
| CoraFluor 1 | |
| 3-hydroxyisobutyryl-CoA hydrolase, EC 3.1.2.4,3-hydroxyisobutyryl-coenzyme A hydrolase3-hydroxyisobutyryl-CoA hydrolase, mitochondrial, HIB-CoA hydrolase, HIBYL-CoA-H, HIBYLCOAH | |
| Full length human recombinant protein of human HIBCH (NP_055177) produced in HEK293T cell. | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 26275 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2a |
| Flow Cytometry, Immunohistochemistry, Western Blot, CyTOF | |
| OTI3H5 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Canine, Human | |
| Purified |