missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Vinpocetine, 98%

Product Code. 15483669
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15483669 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15483669 Supplier Thermo Scientific Chemicals Supplier No. J61000.MC
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specific calcium-calmodulin-dependent phosphodiesterase inhibitor

Specific calcium-calmodulin-dependent phosphodiesterase inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42971-09-5
Molecular Formula C22H26N2O2
Synonym Eburnamenine-14-carboxylic acid ethyl ester

Specifications

Quantity 100 mg
Description White
For Use With (Application) Specific calcium-calmodulin-dependent phosphodiesterase inhibitor
Formula Weight 350.46g/mol
Melting Point 147°C to 149°C
Product Type Signal Transduction Reagent
Form Powder
Assay 98%

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.