missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Violet Red Bile Glucose Agar (ISO)

Product Code. 15829662
Change view
Click to view available options
Fill Weight:
198 to 202 g
99.0-101.0g
Format:
125 mL sirop bottle
250 mL Sirop Bottle
Unit Size:
2 L
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Fill Weight Format unitSize
15829662 99.0-101.0g 125 mL sirop bottle Pack of 10
15795298 198 to 202 g 250 mL Sirop Bottle 2 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15829662 Supplier Thermo Scientific™ Supplier No. BO1329M

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Use this agar for the selective and differential isolation of Enterobacteriaceae

Violet Red Bile Glucose Agar (ISO) is a selective differential medium for the isolation of Enterobacteriaceae

TRUSTED_SUSTAINABILITY

Specifications

Color Dark purple
Content And Storage 2 to 25°C
Format 125 mL sirop bottle
Manufacturing Origin United Kingdom
Product Type Agar
Fill Weight 99.0-101.0g
Packaging Type 10 sirop bottles with screw cap closures in a box.
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.