missing translation for 'onlineSavingsMsg'
Learn More

VIPAS39 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA576148

Additional Details : Weight : 0.01000kg

Product Code. 15942675

  • 516.00€ / 100µL
Estimated Shipment: 24-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

This gene encodes a protein involved in the sorting of lysosomal proteins. Mutations in this gene are associated with ARCS2 (arthrogryposis, renal dysfunction, and cholestasis-2). Alternative splicing results in multiple transcript variants.
Specifications

Specifications

VIPAS39
Polyclonal
Unconjugated
VIPAS39
6720456H09Rik; 9330175H22Rik; AI413782; C10H14orf133; C14orf133; C8H14orf133; hSPE-39; protein spe-39 homolog; SPE39; SPE-39; SPE-39 protein; Spermatogenesis-defective protein 39 homolog; VIPAR; VIPAS39; VPS16B; VPS33B interacting protein, apical-basolateral polarity regulator, spe-39 homolog; VPS33B-interacting protein in apical-basolateral polarity regulator; VPS33B-interacting protein in polarity and apical restriction; VPS33B-interacting protein involved in polarity and apical protein restriction
Rabbit
Antigen affinity chromatography
RUO
104799, 63894, 681989
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q5PQN6, Q8BGQ1, Q9H9C1
VIPAS39
Recombinant protein of human VIPAS39.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions