missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VIPAS39 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA5113490
Additional Details : Weight : 0.01000kg
Description
This gene encodes a protein involved in the sorting of lysosomal proteins. Mutations in this gene are associated with ARCS2 (arthrogryposis, renal dysfunction, and cholestasis-2). Alternative splicing results in multiple transcript variants.Specifications
VIPAS39 | |
Polyclonal | |
Unconjugated | |
VIPAS39 | |
6720456H09Rik; 9330175H22Rik; AI413782; C10H14orf133; C14orf133; C8H14orf133; hSPE-39; protein spe-39 homolog; SPE39; SPE-39; SPE-39 protein; Spermatogenesis-defective protein 39 homolog; VIPAR; VIPAS39; VPS16B; VPS33B interacting protein, apical-basolateral polarity regulator, spe-39 homolog; VPS33B-interacting protein in apical-basolateral polarity regulator; VPS33B-interacting protein in polarity and apical restriction; VPS33B-interacting protein involved in polarity and apical protein restriction | |
Rabbit | |
Protein G | |
RUO | |
104799, 63894 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunocytochemistry, Western Blot | |
3 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q8BGQ1, Q9H9C1 | |
VIPAS39 | |
Recombinant Human Spermatogenesis-defective protein 39 homolog protein (1-493AA) | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |