missing translation for 'onlineSavingsMsg'
Learn More

VIPAS39 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA5120317

Additional Details : Weight : 0.01000kg

Product Code. 17771714

  • 459.00€ / 100µL
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Positive test controls include: Mouse liver, Mouse kidney, Mouse brain, Rat heart, Rat liver. The target is usually found in the following locations: Cytoplasm, Cytoplasmic vesicle, Early endosome, Late endosome, Recycling endosome. Immunogen sequence: MNRTKGDEEE YWNSSKFKAF TFDDEDDELS QLKESKRAVN SLRDFVDDDD DDDLERVSWS GEPVGSISWS IRETAGNSGS THEGREQLKS RNSFSSYAQL PKPTSTYSLS SFFRGRTRPG SFQSLSDALS DTPAKSYAPE LGRPKGEYRD YSNDWSPSDT VRRLRKGKVC SLERFRSLQD KLQLLEEAVS MHDGNVITAV LIFLKRTLSK EILFRELEVR

This gene encodes a protein involved in the sorting of lysosomal proteins. Mutations in this gene are associated with ARCS2 (arthrogryposis, renal dysfunction, and cholestasis-2). Alternative splicing results in multiple transcript variants.
Specifications

Specifications

VIPAS39
Polyclonal
Unconjugated
VIPAS39
6720456H09Rik; 9330175H22Rik; AI413782; C10H14orf133; C14orf133; C8H14orf133; hSPE-39; protein spe-39 homolog; SPE39; SPE-39; SPE-39 protein; Spermatogenesis-defective protein 39 homolog; VIPAR; VIPAS39; VPS16B; VPS33B interacting protein, apical-basolateral polarity regulator, spe-39 homolog; VPS33B-interacting protein in apical-basolateral polarity regulator; VPS33B-interacting protein in polarity and apical restriction; VPS33B-interacting protein involved in polarity and apical protein restriction
Rabbit
Affinity Chromatography
RUO
104799, 681989
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
2.66 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q5PQN6, Q8BGQ1
VIPAS39
Recombinant fusion protein containing a sequence corresponding to amino acids 1-220 of human VIPAS39 (NP_071350.2).
100 μL
Primary
Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions