missing translation for 'onlineSavingsMsg'
Learn More

VIPAS39 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA546768

Additional Details : Weight : 0.00010kg

Product Code. 15597292

  • 472.00€ / 100µL
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Peptide sequence: SDALSDTPAK SYAPELGRPK GEYRDYSNDW SPSDTVRRLR KGKVCSLERF

VIPAS39 Polyclonal antibody specifically detects VIPAS39 in Human samples. It is validated for Western Blot.
Specifications

Specifications

VIPAS39
Polyclonal
Unconjugated
VIPAS39
6720456H09Rik; 9330175H22Rik; AI413782; C10H14orf133; C14orf133; C8H14orf133; hSPE-39; protein spe-39 homolog; SPE39; SPE-39; SPE-39 protein; Spermatogenesis-defective protein 39 homolog; VIPAR; VIPAS39; VPS16B; VPS33B interacting protein, apical-basolateral polarity regulator, spe-39 homolog; VPS33B-interacting protein in apical-basolateral polarity regulator; VPS33B-interacting protein in polarity and apical restriction; VPS33B-interacting protein involved in polarity and apical protein restriction
Rabbit
Affinity Chromatography
RUO
63894
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
Q9H9C1
VIPAS39
Synthetic peptide directed towards the N-terminal of human VIPAS39
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions