Learn More
Viral Dengue Virus 1 NS1 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB94381-SP
This item is not returnable.
View return policy
Description
Dengue Virus NS1 protein Monoclonal specifically detects Dengue Virus NS1 protein in Viral samples. It is validated for ELISA.
Specifications
| Dengue virus NS1 protein | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
| IgG2a |
| ELISA | |
| 973604 | |
| ELISA | |
| P17763 | |
| Human embryonic kidney cell line HEK293-derived viral Dengue Virus 1 NS1 protein, Asp776-Ala1127, Accession No. P17763 | |
| 25 μg | |
| Detects viral Dengue Virus 1 NS1 protein in direct ELISAs. | |
| Virus | |
| Lyophilized |