missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Viral Japanese Encephalitis virus NS1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB10006-100
This item is not returnable.
View return policy
Description
Japanese Encephalitis virus NS1 Monoclonal specifically detects Japanese Encephalitis virus NS1 in Viral samples. It is validated for Western Blot.
Specifications
| Japanese Encephalitis virus NS1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Japanese Encephalitis, JE virus, JEV, JEVgp1, JEV-NS1 | |
| Human embryonic kidney cell, HEK293-derived Japanese Encephalitis virus NS1, Thr796-Ala1146, Accession No. P32886 | |
| 100 μg | |
| 1489713 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG2b |
| Western Blot | |
| 995010 | |
| Western Blot 1 μg;/mL | |
| P32886 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects Japanese Encephalitis virus NS1 in direct ELISAs. | |
| Virus | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction