missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Viral Japanese Encephalitis virus NS1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
193.00€ - 476.00€
Specifications
| Antigen | Japanese Encephalitis virus NS1 |
|---|---|
| Clone | 995047 |
| Dilution | ELISA |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18760737
![]() |
R&D Systems
MAB100061-SP |
25 μg |
193.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18713997
![]() |
R&D Systems
MAB100061-100 |
100 μg |
476.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Japanese Encephalitis virus NS1 Monoclonal specifically detects Japanese Encephalitis virus NS1 in Viral samples. It is validated for ELISA.Specifications
| Japanese Encephalitis virus NS1 | |
| ELISA | |
| Monoclonal | |
| Mouse | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Japanese Encephalitis, JE virus, JEV, JEVgp1, JEV-NS1 | |
| Human embryonic kidney cell, HEK293-derived viral Japanese Encephalitis virus NS1 protein, Thr798-Ala1146, Accession No. P32886 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects viral Japanese Encephalitis virus NS1 protein in direct ELISAs. |
| 995047 | |
| ELISA | |
| Unconjugated | |
| Virus | |
| P32886 | |
| 1489713 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title