Learn More
Leica Microsystems Visoria B Digital Laboratory Microscope for Histopathology or Cytopathology
Product Code. 32177332
Change view
Click to view available options
Magnification Power:
2.5X to 40X
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Magnification Power Unit Size
32177332 2.5X to 40X 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: This product is under eco-contribution tax.
Product Code. 32177332 Supplier Leica Microsystems Supplier No. 111000016
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Regulatory: This product is under eco-contribution tax.
This microscope is suitable for applications in life sciences and clinical laboratories to study histopathology or cytopathology samples. Workflows can be streamlined with encoded functions, optimized light settings, and other features.
  • Visoria B Digital Laboratory Microscope (Global IVD) with illumination management
  • 10 in. screen/tablet
  • Powered by Enersight microscope software platform (Desktop)
  • Flexacam i5 microscope camera, includes power supply
  • High-power white LED
  • Contrast methods: Brightfield
  • Encoded nosepiece, 6x
  • Objectives (N PLAN): 2.5x, 5x, 10x, 20x, 40x
  • ErgoStage with ceramic plate, with specimen holder
  • 2-step focus drive with height-adjustable focus knobs
  • Condenser A 0.9 (P)
  • Dust protection cover
  • Quick start guide and user manual
  • Efficiency through encoded functions
  • More comfort with ergonomics
  • Work without eyepieces by going digital
  • Visualize sample details with the right contrast
TRUSTED_SUSTAINABILITY
Specifications
Eyepiece None
Head Style Monitor
Lighting Type LED
Magnification Power 2.5X to 40X
Model Visoria B
Objective Types Plan
Product Type Biological Microscope