missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VISTA/B7-H5/PD-1H Antibody (CL3975) [DyLight 405], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59030V
This item is not returnable.
View return policy
Description
VISTA/B7-H5/PD-1H Monoclonal antibody specifically detects VISTA/B7-H5/PD-1H in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| VISTA/B7-H5/PD-1H | |
| Monoclonal | |
| DyLight 405 | |
| B7H5, B7-H5, C10orf54, chromosome 10 open reading frame 54, GI24, platelet receptor Gi24, PP2135, SISP1, stress induced secreted protein 1, VSIR | |
| This antibody was developed using a recombinant protein derived from Q9H7M9, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cancer, Cardiovascular Biology, Immunology | |
| 64115 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL3975 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction