missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vitamin D3 Antibody (685503) [DyLight 550], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB6566L
This item is not returnable.
View return policy
Description
Vitamin D3 Monoclonal antibody specifically detects Vitamin D3 in Broad Species Reactivity samples. It is validated for Flow Cytometry (Intracellular Staining), CyTOF
Specifications
| Vitamin D3 | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate | |
| 25-Hydroxyvitamin D3 | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry (Intracellular Staining), CyTOF | |
| 685503 | |
| Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Broad Species Reactivity | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction