Learn More
Vitamin K2(5), TRC
Product Code. 30451252
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30451252 5 mg 5mg
30449876 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30451252 Supplier Toronto Research Chemicals Supplier No. TRCV6761205MG
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 957-78-8
Molecular Formula C16H16O2
Molecular Weight (g/mol) 240.3
Synonym 2-Methyl-3-(3-methyl-2-buten-1-yl)-1,4-naphthalenedione, 2-Methyl-3-(3-methyl-2-butenyl)-1,4-naphthalenedione, 2-Methyl-3-(3-methyl-2-butenyl)-1,4-naphthoquinone, Menaquinone 1, NSC 123509, Vitamin MK 1
IUPAC Name 2-methyl-3-(3-methylbut-2-enyl)naphthalene-1,4-dione
SMILES CC(=CCC1=C(C)C(=O)c2ccccc2C1=O)C
Specifications
Quantity 5 mg
InChI Formula InChI=1S/C16H16O2/c1-10(2)8-9-12-11(3)15(17)13-6-4-5-7-14(13)16(12)18/h4-8H,9H2,1-3H3
Formula Weight 240.115
Chemical Name or Material Vitamin K2(5)