missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vitronectin Monoclonal Antibody (03)
Vitronectin Monoclonal Antibody (3)
Brand: Invitrogen MA531319
This item is not returnable.
View return policy
Description
Specifications
| Vitronectin | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P04004 | |
| VTN | |
| Recombinant Human Vitronectin/VTN Protein. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody |
| ELISA | |
| 03 | |
| Unconjugated | |
| VTN | |
| Aa1018; AI256434; complement S-protein; epibolin; extracellular matrix protein; glycoprotein 66; serum spreading factor; serum-spreading factor; somatomedin B; Somatomedin-B; S-protein; V75; Vitronectin; vitronectin (serum spreading factor, somatomedin B, complement S-protein); Vitronectin V10 subunit; Vitronectin V65 subunit; VN; VNT; VTN | |
| Mouse | |
| Protein A | |
| RUO | |
| 7448 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction