Learn More
Riley™ Vivid Cleaning Spray
Product Code. 31702849
Change view
Click to view available options
Size:
500 mL
Unit Size:
Pack of 25
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
31702849 500 mL Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 31702849 Supplier Riley™ Supplier No. RLYA0006
  • ONLINE EXCLUSIVE PRICE
    192.15€
    160.60€ / Pack of 25
    Save 31.55€
    16% Off
Estimated Shipment: 02-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Convenient cleaning spray for safely and quickly cleaning eyewear lenses, electronic surfaces, and other optical equipment.
  • Anti-reflective
  • Anti-Static
  • Antibacterial
  • Silicone and Alcohol Free
Specifications
Product Type Lens Cleaning Solution
No. per Case 20/Cs.
Size 500 mL
Disposable Limited-use