missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VMAT2 Antibody (CL12531) [mFluor Violet 610 SE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-18570MFV610
This item is not returnable.
View return policy
Description
VMAT2 Monoclonal antibody specifically detects VMAT2 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
Specifications
| VMAT2 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| MGC120477, monoamine neurotransmitter transporter, Monoamine transporter, solute carrier family 18 (vesicular monoamine), member 2, Solute carrier family 18 member 2, SVAT, SVMTMGC120478, synaptic vesicular amine transporter, VAT2MGC26538, vesicle monoamine transporter type 2, vesicle monoamine/H+ antiporter, Vesicular amine transporter 2, VMAT2 | |
| This antibody was developed using a synthetic peptide derived from Q05940, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Neuroscience | |
| 6571 | |
| Store at 4⪚C in the dark. | |
| IgG2b |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CL12531 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction