missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VMAT2 Antibody (CL13157) [Alexa Fluor™ 647], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-18556AF647
This item is not returnable.
View return policy
Description
VMAT2 Monoclonal antibody specifically detects VMAT2 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| VMAT2 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| MGC120477, monoamine neurotransmitter transporter, Monoamine transporter, solute carrier family 18 (vesicular monoamine), member 2, Solute carrier family 18 member 2, SVAT, SVMTMGC120478, synaptic vesicular amine transporter, VAT2MGC26538, vesicle monoamine transporter type 2, vesicle monoamine/H+ antiporter, Vesicular amine transporter 2, VMAT2 | |
| This antibody was developed using a synthetic peptide derived from Q05940, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Neuroscience | |
| 6571 | |
| Store at 4⪚C in the dark. | |
| IgG2b |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL13157 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido