Learn More
VMAT2 Antibody (CL13157) [Allophycocyanin], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-18556APC
Product Code. 30289994
  • 770.00€ / 0.10mL
Description
VMAT2 Monoclonal antibody specifically detects VMAT2 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
VMAT2
Monoclonal
APC
MGC120477, monoamine neurotransmitter transporter, Monoamine transporter, solute carrier family 18 (vesicular monoamine), member 2, Solute carrier family 18 member 2, SVAT, SVMTMGC120478, synaptic vesicular amine transporter, VAT2MGC26538, vesicle monoamine transporter type 2, vesicle monoamine/H+ antiporter, Vesicular amine transporter 2, VMAT2
This antibody was developed using a synthetic peptide derived from Q05940, with the exact immunogen sequence remaining proprietary.
0.1 mL
Neuroscience
6571
Store at 4⪚C in the dark.
IgG2b
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
CL13157
PBS
Mouse
Protein A purified