Learn More
Hirschmann™ Volumetric Measuring Device Fitting
Product Code. 10069770
Change view
Click to view available options
For Use With (Equipment):
Schilling Fast-Flow Burettes, 15 - 50 mL
Schilling Fast-Flow Burettes, 5 - 10 mL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
10069770 Schilling Fast-Flow Burettes, 5 - 10 mL Each
10293861 Schilling Fast-Flow Burettes, 15 - 50 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10069770 Supplier Hirschmann™ Supplier No. 8900710
  • ONLINE EXCLUSIVE PRICE
    24.05€
    22.30€ / Pack of 1
    Save 1.75€
    7% Off
In Stock
Add to basket
Add to basket
Fitting for Dr. Schilling fast-flow burettes
  • For Schilling burettes
Specifications
For Use With (Equipment) Schilling Fast-Flow Burettes, 5 - 10 mL
No. per Pack 1 Each
Type Buret Fitting